HPLC Analysis of Hydrolyzed β-Cyclodextrin on TSKgel® G-Oligo-PW
Materials
analytical column
Product No.
Description
Pricing
TSKgel® G-Oligo-PW HPLC Column
phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle sizeCONDITIONS
column
TSKgel G-Oligo-PW, 30 cm × 7.8 mm I.D., 6 μm particles (808031)
mobile phase
distilled water
flow rate
1 mL/min
column temp.
60 °C
detector
RI
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation