Merck
  • Microwave-assisted synthesis and antimicrobial evaluation of novel spiroisoquinoline and spiropyrido[4,3-d]pyrimidine derivatives.
Microwave-assisted synthesis and antimicrobial evaluation of novel spiroisoquinoline and spiropyrido[4,3-d]pyrimidine derivatives.
Molecules (Basel, Switzerland) (2015-01-28)
Rasha M Faty, Mohamed S Rashed, Mohamed M Youssef
ABSTRACT
Bromination of N-substituted homophthalimides and tetrahydropyrido[4,3-d]- pyrimidine-5,7-diones produces 4,4-dibromohomophthalimide and 8,8-dibromo-tetrahydropyrido[4,3-d]pyrimidine-5,7-dione derivatives, respectively, that can be used as precursors for spiro derivatives. The dibromo derivatives react with different binucleophilic reagents to produce several spiroisoquinoline and spirotetrahydropyrido[4,3-d]- pyrimidine-5,7-dione derivatives, respectively. Reaction of the dibromo derivatives with malononitrile produces dicyanomethylene derivatives which react with different binucleophiles to produce new spiro derivatives. All new compounds are prepared by using the usual chemical conditions and microwave assisted conditions. The latter conditions improved the reaction yields, reduced reaction times and ameliorated the effects on the surrounding environment as the reactions are carried out in closed systems. Structures of the newly synthesized compounds are proved using spectroscopic methods such as IR, MS, 1H-NMR and 13C-NMR and elemental analyses. Some of the newly synthesized compounds were tested for their antimicrobial activities, whereby four of them showed moderate activities and the rest showed low or no activities towards the investigated species.
Product Description
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.00 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 0.18 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm