Merck
  • MiR-129 blocks estrogen induction of NOTCH signaling activity in breast cancer stem-like cells.
MiR-129 blocks estrogen induction of NOTCH signaling activity in breast cancer stem-like cells.
Oncotarget (2017-12-22)
Guodong Xiao, Xiang Li, Gang Li, Boxiang Zhang, Chongwen Xu, Sida Qin, Ning Du, Jichang Wang, Shou-Ching Tang, Jing Zhang, Hong Ren, Ke Chen, Xin Sun
ABSTRACT
Stem-like cells in tumor group featured the major role in the chemotherapy resistance of breast cancer, and the reduction of stem-like cells helped to perish the tumor when receiving chemotherapy. Smaller stem cells number indicated better therapeutic effect in vitro and in clinics, but how did miR-129 and Notch signaling function in breast cancer stem-like cells (BrCSCs) were unclear yet. Through using sphere forming assay and FACS sorting, we found that miR-129 decreased the proportion of stem-like cells in breast cancer cells. Results further indicated that miR-129 degraded the Estrogen Receptor 1 (ESR1) mRNA through a post-translational manner and contributed to the decline of stem-like cells number, preventing tumor regeneration. Cyclin d1 and DICER 1 were proved to promote Let-7 maturation, and in present study, we proved that miR-129 exhibited inhibition on ESR1 and halted the cyclin d1/DICER 1 sustaining of Let-7, which consequently released the Let-7 degradation of NUMB. The restoration of suppressive NUMB by upregulating miR-129 resulted in NOTCH signaling inhibition. In conclusion, we demonstrated the negative regulation of miR-129 on NOTCH signaling activation in BrCSCs' renewal, which was achieved via continuous suppression on cyclin d1/DICER1 sustaining of Let-7 level, and eventually rescued the targeted inhibition of NUMB. The miR-129/ESR1 signaling played pivotal role in controlling DICER1/Let-7/NOTCH cascade via cyclin d1, revealing the novel mechanism of dual Let-7 in non-coding genes network.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCND1
Sigma-Aldrich
MISSION® esiRNA, targeting human ESR1
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
γ Secretase Inhibitor, ≥95% (HPLC)
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Monoclonal Anti-CD24-PE antibody produced in rat, clone SN3, purified immunoglobulin, buffered aqueous solution