Merck
  • Mechanism of radioprotection by dihydroxy-1-selenolane (DHS): Effect of fatty acid conjugation and role of glutathione peroxidase (GPx).
Mechanism of radioprotection by dihydroxy-1-selenolane (DHS): Effect of fatty acid conjugation and role of glutathione peroxidase (GPx).
Biochimie (2017-11-04)
Prachi Verma, Amit Kunwar, Kenta Arai, Michio Iwaoka, K Indira Priyadarsini
ABSTRACT
Dihydroxy-1-selenolane (DHS) previously reported to exhibit radioprotective activity was investigated to understand its mechanism of action in CHO cells of epithelial origin. DHS pre-treatment at 25 μM for 16 h significantly protected CHO cells from radiation (4-11 Gy)-induced delayed mitotic cell death. Further to examine, how increased cellular uptake can influence this mechanism, studies have been performed with DHS-C
Sigma-Aldrich
NU7026, ≥98% (HPLC), solid
Sigma-Aldrich
UCN-01, ≥97% (HPLC), powder
Sigma-Aldrich
CRT0044876, ≥98% (HPLC)
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%