Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
A Pr
A Pr
Physical chemistry chemical physics : PCCP (2017-07-20)
Zheng Gong, Yuxue Liu, Jian Yang, Duanting Yan, Hancheng Zhu, Chunguang Liu, Changshan Xu, Hong Zhang
PMID
28721421
ABSTRACT
Spinel-phase Zn
Product Description
208450
Sigma-Aldrich
Germanium(IV) chloride, 99.99% trace metals basis
Pricing
289892
Sigma-Aldrich
Gallium(III) nitrate hydrate, crystalline, 99.9% trace metals basis
Pricing