Merck
  • The α2β2 isoform combination dominates the astrocytic Na+ /K+ -ATPase activity and is rendered nonfunctional by the α2.G301R familial hemiplegic migraine type 2-associated mutation.
The α2β2 isoform combination dominates the astrocytic Na+ /K+ -ATPase activity and is rendered nonfunctional by the α2.G301R familial hemiplegic migraine type 2-associated mutation.
Glia (2017-08-09)
Anca Stoica, Brian Roland Larsen, Mette Assentoft, Rikke Holm, Leanne Melissa Holt, Frederik Vilhardt, Bente Vilsen, Karin Lykke-Hartmann, Michelle Lynne Olsen, Nanna MacAulay
ABSTRACT
Synaptic activity results in transient elevations in extracellular K+ , clearance of which is critical for sustained function of the nervous system. The K+ clearance is, in part, accomplished by the neighboring astrocytes by mechanisms involving the Na+ /K+ -ATPase. The Na+ /K+ -ATPase consists of an α and a β subunit, each with several isoforms present in the central nervous system, of which the α2β2 and α2β1 isoform combinations are kinetically geared for astrocytic K+ clearance. While transcript analysis data designate α2β2 as predominantly astrocytic, the relative quantitative protein distribution and isoform pairing remain unknown. As cultured astrocytes altered their isoform expression in vitro, we isolated a pure astrocytic fraction from rat brain by a novel immunomagnetic separation approach in order to determine the expression levels of α and β isoforms by immunoblotting. In order to compare the abundance of isoforms in astrocytic samples, semi-quantification was carried out with polyhistidine-tagged Na+ /K+ -ATPase subunit isoforms expressed in Xenopus laevis oocytes as standards to obtain an efficiency factor for each antibody. Proximity ligation assay illustrated that α2 paired efficiently with both β1 and β2 and the semi-quantification of the astrocytic fraction indicated that the astrocytic Na+ /K+ -ATPase is dominated by α2, paired with β1 or β2 (in a 1:9 ratio). We demonstrate that while the familial hemiplegic migraine-associated α2.G301R mutant was not functionally expressed at the plasma membrane in a heterologous expression system, α2+/G301R mice displayed normal protein levels of α2 and glutamate transporters and that the one functional allele suffices to manage the general K+ dynamics.
Sigma-Aldrich
Anti-ATP1B2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Glutamate Transporter Antibody, Glial, serum, Chemicon®
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Na+K+ ATPase α-2 Antibody, serum, Upstate®