Merck
  • Congener-specific determination of ultratrace levels of chlorinated and brominated polycyclic aromatic hydrocarbons in atmosphere and industrial stack gas by isotopic dilution gas chromatography/high resolution mass spectrometry method.
Congener-specific determination of ultratrace levels of chlorinated and brominated polycyclic aromatic hydrocarbons in atmosphere and industrial stack gas by isotopic dilution gas chromatography/high resolution mass spectrometry method.
Journal of chromatography. A (2017-06-18)
Rong Jin, Guorui Liu, Minghui Zheng, Heidelore Fiedler, Xiaoxu Jiang, Lili Yang, Xiaolin Wu, Yang Xu
ABSTRACT
Isotopic dilution gas chromatography combined with high resolution mass spectrometry (GC/HRMS) has overwhelming advantages with respect to the accuracy of congener-specific ultratrace analysis of complex persistent organic pollutants (POPs) in environmental matrices. However, an isotopic dilution GC/HRMS method for analysis of chlorinated and brominated polycyclic aromatic hydrocarbons (Cl-PAHs and Br-PAHs) using
Sigma-Aldrich
3-Bromophenanthrene, 96%
Sigma-Aldrich
9,10-Dibromoanthracene, 98%
Sigma-Aldrich
9-Bromophenanthrene, 96%
Sigma-Aldrich
2-Bromofluorene, 95%
Sigma-Aldrich
5-Bromoacenaphthene
Sigma-Aldrich
9-Chlorofluorene, AldrichCPR
Sigma-Aldrich
9-Bromoanthracene, 94%
Sigma-Aldrich
1-Bromopyrene, 96%
Sigma-Aldrich
2,7-Dibromofluorene, 97%