Merck
  • Expression, purification and use of recombinant annexin V for the detection of apoptotic cells.
Expression, purification and use of recombinant annexin V for the detection of apoptotic cells.
Nature protocols (2009-09-05)
Susan E Logue, Mohamed Elgendy, Seamus J Martin
ABSTRACT
Apoptosis is a mode of programmed cell death that is widely used to eliminate cells during development, tissue homeostasis, infection or in response to injury. Alterations to the plasma membranes of apoptotic cells trigger recognition and engulfment of such cells by phagocytes. Measurement of plasma membrane phosphatidylserine externalization, using fluorescently labeled annexin V, is widely used for the detection of apoptotic cells. Here we describe protocols for bacterial expression, purification and FITC labeling of recombinant annexin V. By following the method outlined in this protocol, it is possible to produce milligram amounts of recombinant annexin V within 3 d. We also describe a method for the assessment of annexin V binding to cell populations by flow cytometry or fluorescence microscopy.
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Leupeptin, >4000 U/mg
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%