Merck
  • The metabolic function of cyclin D3-CDK6 kinase in cancer cell survival.
The metabolic function of cyclin D3-CDK6 kinase in cancer cell survival.
Nature (2017-06-14)
Haizhen Wang, Brandon N Nicolay, Joel M Chick, Xueliang Gao, Yan Geng, Hong Ren, Hui Gao, Guizhi Yang, Juliet A Williams, Jan M Suski, Mark A Keibler, Ewa Sicinska, Ulrike Gerdemann, W Nicholas Haining, Thomas M Roberts, Kornelia Polyak, Steven P Gygi, Nicholas J Dyson, Piotr Sicinski
ABSTRACT
D-type cyclins (D1, D2 and D3) and their associated cyclin-dependent kinases (CDK4 and CDK6) are components of the core cell cycle machinery that drives cell proliferation. Inhibitors of CDK4 and CDK6 are currently being tested in clinical trials for patients with several cancer types, with promising results. Here, using human cancer cells and patient-derived xenografts in mice, we show that the cyclin D3-CDK6 kinase phosphorylates and inhibits the catalytic activity of two key enzymes in the glycolytic pathway, 6-phosphofructokinase and pyruvate kinase M2. This re-directs the glycolytic intermediates into the pentose phosphate (PPP) and serine pathways. Inhibition of cyclin D3-CDK6 in tumour cells reduces flow through the PPP and serine pathways, thereby depleting the antioxidants NADPH and glutathione. This, in turn, increases the levels of reactive oxygen species and causes apoptosis of tumour cells. The pro-survival function of cyclin D-associated kinase operates in tumours expressing high levels of cyclin D3-CDK6 complexes. We propose that measuring the levels of cyclin D3-CDK6 in human cancers might help to identify tumour subsets that undergo cell death and tumour regression upon inhibition of CDK4 and CDK6. Cyclin D3-CDK6, through its ability to link cell cycle and cell metabolism, represents a particularly powerful oncoprotein that affects cancer cells at several levels, and this property can be exploited for anti-cancer therapy.
Sigma-Aldrich
CDK6/CyclinD1, active, His/GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
2-Aminopyridine-3-carboxamide, 97%
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Anti-BRCA1 Antibody, serum, Upstate®
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, clone MPM-2, Upstate®, from mouse