- Mapping Polymer Donors toward High-Efficiency Fullerene Free Organic Solar Cells.
Mapping Polymer Donors toward High-Efficiency Fullerene Free Organic Solar Cells.
Advanced materials (Deerfield Beach, Fla.) (2016-11-20)
Yuze Lin, Fuwen Zhao, Yang Wu, Kai Chen, Yuxin Xia, Guangwu Li, Shyamal K K Prasad, Jingshuai Zhu, Lijun Huo, Haijun Bin, Zhi-Guo Zhang, Xia Guo, Maojie Zhang, Yanming Sun, Feng Gao, Zhixiang Wei, Wei Ma, Chunru Wang, Justin Hodgkiss, Zhishan Bo, Olle Inganäs, Yongfang Li, Xiaowei Zhan
PMID27862373
ABSTRACT
Five polymer donors with distinct chemical structures and different electronic properties are surveyed in a planar and narrow-bandgap fused-ring electron acceptor (IDIC)-based organic solar cells, which exhibit power conversion efficiencies of up to 11%.