Merck
  • Efficient and Air-Stable Mixed-Cation Lead Mixed-Halide Perovskite Solar Cells with n-Doped Organic Electron Extraction Layers.
Efficient and Air-Stable Mixed-Cation Lead Mixed-Halide Perovskite Solar Cells with n-Doped Organic Electron Extraction Layers.
Advanced materials (Deerfield Beach, Fla.) (2016-12-03)
Zhiping Wang, David P McMeekin, Nobuya Sakai, Stephan van Reenen, Konrad Wojciechowski, Jay B Patel, Michael B Johnston, Henry J Snaith
ABSTRACT
Air-stable doping of the n-type fullerene layer in an n-i-p planar heterojunction perovskite device is capable of enhancing device efficiency and improving device stability. Employing a (HC(NH
Sigma-Aldrich
Dimethylammonium bromide, ≥98%
Sigma-Aldrich
Formamidinium iodide, ≥99%, anhydrous
Sigma-Aldrich
Methylammonium bromide, ≥99%, anhydrous
Sigma-Aldrich
Formamidinium bromide, ≥99%, anhydrous
Sigma-Aldrich
Imidazolium bromide, ≥98%
Sigma-Aldrich
Methylammonium iodide, ≥99%, anhydrous
Sigma-Aldrich
Formamidinium bromide, ≥98%
Sigma-Aldrich
Guanidinium bromide, ≥98%
Sigma-Aldrich
Ethane-1,2-diammonium bromide, ≥98%
Sigma-Aldrich
Ethane-1,2-diammonium iodide, ≥98%
Sigma-Aldrich
Guanidinium iodide, ≥99%, anhydrous
Sigma-Aldrich
Phenethylammonium bromide, ≥98%
Sigma-Aldrich
n-Butylammonium bromide, ≥98%