Merck
  • High-Performance Electron Acceptor with Thienyl Side Chains for Organic Photovoltaics.
High-Performance Electron Acceptor with Thienyl Side Chains for Organic Photovoltaics.
Journal of the American Chemical Society (2016-03-26)
Yuze Lin, Fuwen Zhao, Qiao He, Lijun Huo, Yang Wu, Timothy C Parker, Wei Ma, Yanming Sun, Chunru Wang, Daoben Zhu, Alan J Heeger, Seth R Marder, Xiaowei Zhan
ABSTRACT
We develop an efficient fused-ring electron acceptor (ITIC-Th) based on indacenodithieno[3,2-b]thiophene core and thienyl side-chains for organic solar cells (OSCs). Relative to its counterpart with phenyl side-chains (ITIC), ITIC-Th shows lower energy levels (ITIC-Th: HOMO = -5.66 eV, LUMO = -3.93 eV; ITIC: HOMO = -5.48 eV, LUMO = -3.83 eV) due to the σ-inductive effect of thienyl side-chains, which can match with high-performance narrow-band-gap polymer donors and wide-band-gap polymer donors. ITIC-Th has higher electron mobility (6.1 × 10(-4) cm(2) V(-1) s(-1)) than ITIC (2.6 × 10(-4) cm(2) V(-1) s(-1)) due to enhanced intermolecular interaction induced by sulfur-sulfur interaction. We fabricate OSCs by blending ITIC-Th acceptor with two different low-band-gap and wide-band-gap polymer donors. In one case, a power conversion efficiency of 9.6% was observed, which rivals some of the highest efficiencies for single junction OSCs based on fullerene acceptors.
Sigma-Aldrich
ITIC