Merck
  • Application of Ionic Liquids to Energy Storage and Conversion Materials and Devices.
Application of Ionic Liquids to Energy Storage and Conversion Materials and Devices.
Chemical reviews (2017-01-14)
Masayoshi Watanabe, Morgan L Thomas, Shiguo Zhang, Kazuhide Ueno, Tomohiro Yasuda, Kaoru Dokko
ABSTRACT
Ionic liquids (ILs) are liquids consisting entirely of ions and can be further defined as molten salts having melting points lower than 100 °C. One of the most important research areas for IL utilization is undoubtedly their energy application, especially for energy storage and conversion materials and devices, because there is a continuously increasing demand for clean and sustainable energy. In this article, various application of ILs are reviewed by focusing on their use as electrolyte materials for Li/Na ion batteries, Li-sulfur batteries, Li-oxygen batteries, and nonhumidified fuel cells and as carbon precursors for electrode catalysts of fuel cells and electrode materials for batteries and supercapacitors. Due to their characteristic properties such as nonvolatility, high thermal stability, and high ionic conductivity, ILs appear to meet the rigorous demands/criteria of these various applications. However, for further development, specific applications for which these characteristic properties become unique (i.e., not easily achieved by other materials) must be explored. Thus, through strong demands for research and consideration of ILs unique properties, we will be able to identify indispensable applications for ILs.
Sigma-Aldrich
1-Butyl-2,3-dimethylimidazolium bis(trifluoromethylsulfonyl)imide, ≥99%, H2O <500 ppm
Sigma-Aldrich
1-Ethyl-3-methylimidazolium tetrafluoroborate, ≥99%, <1000 ppm H2O
Sigma-Aldrich
1-Methyl-1-propylpiperidinium bis(trifluoromethylsulfonyl)imide, ≥99%, H2O ≤500 ppm
Sigma-Aldrich
1-Ethyl-3-methylimidazolium chloride, >99%
Sigma-Aldrich
1-Ethyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, ≥99%, H2O <500 ppm
Sigma-Aldrich
1-Ethyl-3-methylimidazolium acetate, ≥98%
Sigma-Aldrich
1-Butyl-3-methylimidazolium chloride, ≥99%
Sigma-Aldrich
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, >99%, <500 ppm H2O
Sigma-Aldrich
1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, ≥99%, H2O ≤500 ppm
Sigma-Aldrich
1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, ≥99%, H2O <500 ppm
Sigma-Aldrich
1-Ethyl-3-methylimidazolium hexafluorophosphate, ≥99%, <500 ppm H2O