Merck
  • The progression of CD56+ myeloid sarcoma: A case report and literature review.
The progression of CD56+ myeloid sarcoma: A case report and literature review.
Oncology letters (2016-04-29)
Xin Wang, Wen-Sheng Li, Yan Zheng, Zhao-Xia Ying, Yong-Xian Wang, Ying-Mei Wang, Jun-Feng Zheng, Sheng-Xiang Xiao
ABSTRACT
The current study presents a case of cluster of differentiation (CD)56+ myeloid sarcoma in a patient that initially presented with skin lesions, and provides evidence for the clinical and differential diagnosis of myeloid sarcoma. The patient of the present case report was a 65-year-old man who was admitted to hospital with a six-month history of bilateral purple-red papules and nodules, which were present on the upper limbs of the patient and had spread over his whole body one month prior to admission to the hospital. Pathological examination demonstrated a diffuse infusion of primitive round cells at the papillary dermis and subcutaneous tissues. The infiltrated cells were 40-60 µm in diameter and morphologically identical. Immunohistochemical examination revealed that the cells expressed myeloperoxidase, CD56, CD43 and T-cell intracytoplasmic antigen. In addition, several cells expressed CD34, and 90% of the cells expressed Ki67. While the majority of cells in myeloid sarcoma do not express CD56, the present case was a myeloid sarcoma that expressed CD56, which is extremely rare. The sarcoma in the present patient progressed rapidly, and the patient died eight months following the onset of disease. Clinicians should be aware of CD56+ myeloid sarcoma, which is easily misdiagnosed and inappropriately treated. Consequently, myeloid sarcoma may have a high malignancy and poor outcome for patients.
Sigma-Aldrich
Monoclonal Anti-Proliferating Cell Protein Ki-67 antibody produced in mouse, clone PP-67, ascites fluid
Sigma-Aldrich
Anti-PAX-5 antibody, Rabbit monoclonal, clone SP34, recombinant, expressed in proprietary host, affinity isolated antibody
Sigma-Aldrich
Anti-CD20 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP32, affinity isolated antibody
Sigma-Aldrich
Anti-GZMB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MPO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-CD34 antibody produced in mouse, clone 581, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CD7 antibody, Rabbit monoclonal, clone SP94, recombinant, expressed in proprietary host, affinity isolated antibody
Sigma-Aldrich
Anti-CD3 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP162, affinity isolated antibody
Sigma-Aldrich
Anti-CD4 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP35, affinity isolated antibody