Merck
  • Drug safety evaluation of oral tenofovir disoproxil fumarate-emtricitabine for pre-exposure prophylaxis for human immunodeficiency virus infection.
Drug safety evaluation of oral tenofovir disoproxil fumarate-emtricitabine for pre-exposure prophylaxis for human immunodeficiency virus infection.
Expert opinion on drug safety (2016-07-09)
Tracy P Trang, Betty J Dong, Noah Kojima, Jeffrey D Klausner
ABSTRACT
Tenofovir disoproxil fumarate (TDF) and emtricitabine (FTC) are nucleoside reverse transcriptase inhibitors approved as pre-exposure prophylaxis (PrEP) against human immunodeficiency virus (HIV). Prophylactic TDF-based regimens have been shown to reduce the risk of HIV infection by 74 to 92% among participants with detectable drug levels. Adverse events observed in clinical trials include nausea, elevated creatinine and liver enzymes, and decreased bone mineral density. This article reviews the pharmacology, pharmacokinetics, and the safety profile of TDF and FTC used as PrEP for HIV infection. TDF-FTC can have a large impact in preventing HIV infections among high risk individuals when taken daily. Although TDF-FTC is associated with adverse events, they can be minimized with clinician-guided monitoring.
Sigma-Aldrich
Tenofovir, ≥98% (HPLC)
Sigma-Aldrich
Tenofovir disoproxil fumarate, ≥98% (HPLC)