Merck
  • Allelic series of Huntington's disease knock-in mice reveals expression discorrelates.
Allelic series of Huntington's disease knock-in mice reveals expression discorrelates.
Human molecular genetics (2016-02-26)
Ashish Kumar, Jennifer Zhang, Sara Tallaksen-Greene, Michael R Crowley, David K Crossman, A Jennifer Morton, Thomas Van Groen, Inga Kadish, Roger L Albin, Mathieu Lesort, Peter J Detloff
ABSTRACT
Identifying molecular drivers of pathology provides potential therapeutic targets. Differentiating between drivers and coincidental molecular alterations presents a major challenge. Variation unrelated to pathology further complicates transcriptomic, proteomic and metabolomic studies which measure large numbers of individual molecules. To overcome these challenges towards the goal of determining drivers of Huntington's disease (HD), we generated an allelic series of HD knock-in mice with graded levels of phenotypic severity for comparison with molecular alterations. RNA-sequencing analysis of this series reveals high numbers of transcripts with level alterations that do not correlate with phenotypic severity. These discorrelated molecular changes are unlikely to be drivers of pathology allowing an exclusion-based strategy to provide a short list of driver candidates. Further analysis of the data shows that a majority of transcript level changes in HD knock-in mice involve alteration of the rate of mRNA processing and/or degradation rather than solely being due to alteration of transcription rate. The overall strategy described can be applied to assess the influence of any molecular change on pathology for diseases where different mutations cause graded phenotypic severity.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, ascites fluid, clone 5TF1-1C2, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Pricing and availability is not currently available.