Merck
  • SNP haplotyping technique for evaluation of MGP 5' UTR power in osteoblast cells.
SNP haplotyping technique for evaluation of MGP 5' UTR power in osteoblast cells.
SpringerPlus (2016-07-05)
Abazar Roustazadeh, Seyed Hamidreza Monavari, Seyed Reza Hosseini Fard, Parisa Hassanpour, Amaneh Yarnazari, Mohammad Najafi
ABSTRACT
Matrix Gla protein (MGP) is involved in calcium trafficking and arterial calcification. The aim of study was to investigate the role of three polymorphisms within the MGP gene promoter region on reporter gene (luciferase) expression level. The fragments containing rs1800799 (C/T), rs1800802 (T/C), and rs1800801 (G/A) sites were constructed and transferred into human G292 osteoblast cells using pGL3-Basic plasmid. The reporter gene expression was calculated for the high and low frequency polymorphic haplotypes (CTG and TCA, respectively). Results showed that the reporter gene expression levels are not statistically different (p > 0.3). We concluded that the investigated polymorphic sites are not able to change the gene expression pattern in human G292 osteoblast cells.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines