Merck
  • PRMT1 promotes mitosis of cancer cells through arginine methylation of INCENP.
PRMT1 promotes mitosis of cancer cells through arginine methylation of INCENP.
Oncotarget (2015-10-16)
Xiaolan Deng, Gottfried Von Keudell, Takehiro Suzuki, Naoshi Dohmae, Makoto Nakakido, Lianhua Piao, Yuichiro Yoshioka, Yusuke Nakamura, Ryuji Hamamoto
ABSTRACT
Inner centromere protein (INCENP) is a part of a protein complex known as the chromosomal passenger complex (CPC) that is essential for correcting non-bipolar chromosome attachments and for cytokinesis. We here demonstrate that a protein arginine methyltransferase PRMT1, which are overexpressed in various types of cancer including lung and bladder cancer, methylates arginine 887 in an Aurora Kinase B (AURKB)-binding region of INCENP both in vitro and in vivo. R887-substituted INCENP revealed lower binding-affinity to AURKB than wild-type INCENP in the presence of PRMT1. Knockdown of PRMT1 as well as overexpression of methylation-inactive INCENP attenuated the AURKB activity in cancer cells, and resulted in abnormal chromosomal alignment and segregation. Furthermore, introduction of methylation-inactive INCENP into cancer cells reduced the growth rate, compared with those introduced wild-type INCENP or Mock. Our data unveils a novel mechanism of PRMT1-mediated CPC regulation through methylation of INCENP.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
PRMT1 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
PRMT1 human, recombinant, expressed in E. coli, ≥50% (SDS-PAGE)
Sigma-Aldrich
PRMT1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O