Merck
Identification of TAX2 peptide as a new unpredicted anti-cancer agent.
Oncotarget (2015-06-06)
Albin Jeanne, Emilie Sick, Jérôme Devy, Nicolas Floquet, Nicolas Belloy, Louis Theret, Camille Boulagnon-Rombi, Marie-Danièle Diebold, Manuel Dauchez, Laurent Martiny, Christophe Schneider, Stéphane Dedieu
ABSTRACT
The multi-modular glycoprotein thrombospondin-1 (TSP-1) is considered as a key actor within the tumor microenvironment. Besides, TSP-1 binding to CD47 is widely reported to regulate cardiovascular function as it promotes vasoconstriction and angiogenesis limitation. Therefore, many studies focused on targeting TSP-1:CD47 interaction, aiming for up-regulation of physiological angiogenesis to enhance post-ischemia recovery or to facilitate engraftment. Thus, we sought to identify an innovative selective antagonist for TSP-1:CD47 interaction. Protein-protein docking and molecular dynamics simulations were conducted to design a novel CD47-derived peptide, called TAX2. TAX2 binds TSP-1 to prevent TSP-1:CD47 interaction, as revealed by ELISA and co-immunoprecipitation experiments. Unexpectedly, TAX2 inhibits in vitro and ex vivo angiogenesis features in a TSP-1-dependent manner. Consistently, our data highlighted that TAX2 promotes TSP-1 binding to CD36-containing complexes, leading to disruption of VEGFR2 activation and downstream NO signaling. Such unpredicted results prompted us to investigate TAX2 potential in tumor pathology. A multimodal imaging approach was conducted combining histopathological staining, MVD, MRI analysis and μCT monitoring for tumor angiography longitudinal follow-up and 3D quantification. TAX2 in vivo administrations highly disturb syngeneic melanoma tumor vascularization inducing extensive tumor necrosis and strongly inhibit growth rate and vascularization of human pancreatic carcinoma xenografts in nude mice.
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Anti-LRP, Heavy Chain Mouse mAb (8G1), liquid, clone 8G1, Calbiochem®
Sigma-Aldrich
MISSION® esiRNA, targeting human GZMA
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Thbs1
Sigma-Aldrich
MISSION® esiRNA, targeting human THBS1
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder
Supelco
Cannabidiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®