Merck
  • Synthesis, characterization, photophysics and electrochemical study of luminescent iridium(III) complexes with isocyanoborate ligands.
Synthesis, characterization, photophysics and electrochemical study of luminescent iridium(III) complexes with isocyanoborate ligands.
Dalton transactions (Cambridge, England : 2003) (2015-05-06)
Kin-Cheung Chan, Wing-Kin Chu, Shek-Man Yiu, Chi-Chiu Ko
ABSTRACT
A new series of cyclometalated iridium(III) complexes with isocyanoborate ligands [Ir(R2ppy)2(L)(CNBR'3)] (R = H or F; L = CNC6H4Cl-4 or PPh3; R' = Ph, C6F5 or C6H4Cl-4), [Ir(biqb)(ppy)(CNBR''3)] (R'' = C6F5 or C6H4Cl-4) and {Ir(ppy)2(CN)n[CNB(C6F5)3]2-n}(-) (n = 0 or 1) have been synthesized and characterized. Three of these complexes have also been structurally characterized by X-ray crystallography. The photophysical and electrochemical properties of these complexes have been investigated. The effects of isocyanoborate ligands on the luminescence properties of these iridium(III) complexes are also described.
Sigma-Aldrich
2-Phenylpyridine, 98%
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
2-(2,4-Difluorophenyl)pyridine, 97%
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
Boron-11B, 95 atom % 11B
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)