Merck
Synthesis of deuterium-labelled isotopomer of deferasirox.
Journal of labelled compounds & radiopharmaceuticals (2015-02-19)
Freddy H Havaldar, Bhushan Vasant Dabholkar, Ganesh Baban Mule, Suhas Kulkarni
ABSTRACT
A d4 -labeled isotopomer of deferasirox was synthesized as internal standard for use in a LC/mass spectroscopy (MS)/MS method developed for the simultaneous quantitative determination of deferasirox in human serum. d4 -deferasirox was synthesized from d8 -toluene.
Sigma-Aldrich
Toluene-d8, 99 atom % D
Sigma-Aldrich
Toluene-d8, 99 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Toluene-d8, "100%", 99.96 atom % D
Sigma-Aldrich
Toluene-d8, 99.6 atom % D