Merck
  • [Au5Mes5]: improved gram-scale synthesis and its use as a convenient precursor for halide-free supported gold nanoparticles.
[Au5Mes5]: improved gram-scale synthesis and its use as a convenient precursor for halide-free supported gold nanoparticles.
Dalton transactions (Cambridge, England : 2003) (2015-07-23)
G Siddiqi, V Mougel, C Copéret
ABSTRACT
A simple one-step and gram-scale synthesis of [Au5Mes5] from AuCl3 was developed, and this molecular precursor was used to generate Au nanoparticles on SiO2 and Al2O3. While [Au5Mes5] does not react with surface silanols and is only physisorbed, its incipient wetness impregnation followed by H2 treatment leads to a narrow size distribution of 1.4 nm Au nanoparticles. In contrast, [Au5Mes5] reacts with partially dehydroxylated Al2O3 to directly yield 1 nm Au nanoparticles along with adsorbed species. A subsequent treatment under hydrogen leads to a narrow size distribution of smaller 0.8 nm Au particles.
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O