Merck
  • Extension of a dynamic headspace multi-volatile method to milliliter injection volumes with full sample evaporation: Application to green tea.
Extension of a dynamic headspace multi-volatile method to milliliter injection volumes with full sample evaporation: Application to green tea.
Journal of chromatography. A (2015-08-16)
Nobuo Ochiai, Kikuo Sasamoto, Jun Tsunokawa, Andreas Hoffmann, Kazunori Okanoya, Kevin MacNamara
ABSTRACT
An extension of multi-volatile method (MVM) technology using the combination of a standard dynamic headspace (DHS) configuration, and a modified DHS configuration incorporating an additional vacuum module, was developed for milliliter injection volume of aqueous sample with full sample evaporation. A prior step involved investigation of water management by weighing of the water residue in the adsorbent trap. The extended MVM for 1 mL aqueous sample consists of five different DHS method parameter sets including choice of the replaceable adsorbent trap. An initial two DHS sampling sets at 25°C with the standard DHS configuration using a carbon-based adsorbent trap target very volatile solutes with high vapor pressure (>10 kPa) and volatile solutes with moderate vapor pressure (1-10 kPa). Subsequent three DHS sampling sets at 80°C with the modified DHS configuration using a Tenax TA trap target solutes with low vapor pressure (<1 kPa) and/or hydrophilic characteristics. After the five sequential DHS samplings using the same HS vial, the five traps are sequentially desorbed with thermal desorption in reverse order of the DHS sampling and the desorbed compounds are trapped and concentrated in a programmed temperature vaporizing (PTV) inlet and subsequently analyzed in a single GC-MS run. Recoveries of 21 test aroma compounds in 1 mL water for each separate DHS sampling and the combined MVM procedure were evaluated as a function of vapor pressure in the range of 0.000088-120 kPa. The MVM procedure provided high recoveries (>88%) for 17 test aroma compounds and moderate recoveries (44-71%) for 4 test compounds. The method showed good linearity (r(2)>0.9913) and high sensitivity (limit of detection: 0.1-0.5 ng mL(-1)) even with MS scan mode. The improved sensitivity of the method was demonstrated with analysis of a wide variety of aroma compounds in brewed green tea. Compared to the original 100 μL MVM procedure, this extension to 1 mL MVM allowed detection of nearly twice the number of aroma compounds, including 18 potent aroma compounds from top-note to base-note (e.g. 2,3-butanedione, coumarin, furaneol, guaiacol, cis-3-hexenol, linalool, maltol, methional, 3-methyl butanal, 2,3,5-trimethyl pyrazine, and vanillin). Sensitivity for 23 compounds improved by a factor of 3.4-15 under 1 mL MVM conditions.
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
2,6-Dimethylpyrazine, 98%
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Butyraldehyde, ≥96.0%
Sigma-Aldrich
Butyraldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Butyraldehyde, natural, FG
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Furan, ≥99%
Sigma-Aldrich
2,3-Pentanedione, 97%
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
2,6-Dimethylpyrazine, ≥98%, FG
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
2,3-Butanedione, 97%
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Valeraldehyde, 97%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
2,3-Pentanedione, natural, ≥96%, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
2,3-Pentanedione, ≥96%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Methional, ≥97%, FG
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG