Merck
  • The androgen receptor controls expression of the cancer-associated sTn antigen and cell adhesion through induction of ST6GalNAc1 in prostate cancer.
The androgen receptor controls expression of the cancer-associated sTn antigen and cell adhesion through induction of ST6GalNAc1 in prostate cancer.
Oncotarget (2015-10-10)
Jennifer Munkley, Sebastian Oltean, Daniel Vodák, Brian T Wilson, Karen E Livermore, Yan Zhou, Eleanor Star, Vasileios I Floros, Bjarne Johannessen, Bridget Knight, Paul McCullagh, John McGrath, Malcolm Crundwell, Rolf I Skotheim, Craig N Robson, Hing Y Leung, Lorna W Harries, Prabhakar Rajan, Ian G Mills, David J Elliott
ABSTRACT
Patterns of glycosylation are important in cancer, but the molecular mechanisms that drive changes are often poorly understood. The androgen receptor drives prostate cancer (PCa) development and progression to lethal metastatic castration-resistant disease. Here we used RNA-Seq coupled with bioinformatic analyses of androgen-receptor (AR) binding sites and clinical PCa expression array data to identify ST6GalNAc1 as a direct and rapidly activated target gene of the AR in PCa cells. ST6GalNAc1 encodes a sialytransferase that catalyses formation of the cancer-associated sialyl-Tn antigen (sTn), which we find is also induced by androgen exposure. Androgens induce expression of a novel splice variant of the ST6GalNAc1 protein in PCa cells. This splice variant encodes a shorter protein isoform that is still fully functional as a sialyltransferase and able to induce expression of the sTn-antigen. Surprisingly, given its high expression in tumours, stable expression of ST6GalNAc1 in PCa cells reduced formation of stable tumours in mice, reduced cell adhesion and induced a switch towards a more mesenchymal-like cell phenotype in vitro. ST6GalNAc1 has a dynamic expression pattern in clinical datasets, beingsignificantly up-regulated in primary prostate carcinoma but relatively down-regulated in established metastatic tissue. ST6GalNAc1 is frequently upregulated concurrently with another important glycosylation enzyme GCNT1 previously associated with prostate cancer progression and implicated in Sialyl Lewis X antigen synthesis. Together our data establishes an androgen-dependent mechanism for sTn antigen expression in PCa, and are consistent with a general role for the androgen receptor in driving important coordinate changes to the glycoproteome during PCa progression.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Arhgap4
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b8
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt, ≥85% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human ARHGAP4
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human AREG
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting human AKR1B1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fdxr
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Areg
Sigma-Aldrich
MISSION® esiRNA, targeting human AR
Sigma-Aldrich
MISSION® esiRNA, targeting human FDXR
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Anti-ST6GALNAC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution