Merck
Analysis of adipose tissue lipid using mass spectrometry.
Methods in enzymology (2014-02-18)
Rodney C Baker, Yana Nikitina, Angela R Subauste
ABSTRACT
Mass spectrometry technology has enabled significant advances in detailing the alterations of the lipidome in response to pathological conditions or experimental manipulations. Lipids comprise a wide range of compounds with functions that include structural, intracellular signaling, trafficking, and storage. Characterization of lipid species has evolved significantly over recent years due to the progress made in the area of mass spectrometry. This chapter details the methods used for the analysis of lipids tailored to the intrinsic characteristics of adipose tissue. Particular attention is given to the analysis of triglycerides, diacylglycerols, and phospholipid.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer