Merck
Resveratrol transport and metabolism by human intestinal Caco-2 cells.
The Journal of pharmacy and pharmacology (2003-05-02)
Mark I Kaldas, U Kristina Walle, Thomas Walle
ABSTRACT
Resveratrol is a dietary constituent suggested to have protective effects against cancer as well as cardiovascular disease. The purpose of the study was to learn whether this agent could be absorbed in man and enter the systemic circulation. This was examined by measuring transport and metabolism of resveratrol (5-40 microM) by the human intestinal epithelial cell line Caco-2 cultured in Transwells. Transport across the Caco-2 monolayer occurred in a direction-independent manner with P(app) values of approximately 7 x 10(-6) cm s(-1), much higher than for the paracellular transport marker mannitol (approximately 0.4 x 10(-6) cm s(-1)), suggesting efficient absorption in-vivo. At the highest resveratrol concentration, the absorption increased, possibly due to saturation of metabolism. In sharp contrast to previous findings in the rat, the metabolism of resveratrol in Caco-2 cells involved mainly sulfation and, to a minor extent, glucuronidation. At low resveratrol concentrations, most of the sulfate conjugate was exported to the apical side, presumably by MRP2, which is well expressed in these cells. At high concentrations, there was a shift towards the basolateral side, possibly involving MRP3, which was recently shown also to be expressed in Caco-2 cells. These results indicate that absorption of resveratrol in-vivo may be high but with limited bioavailability due to efficient sulfate conjugation. Extensive accumulation of resveratrol in the Caco-2 cells, demonstrated in additional experiments, suggests enterocytes as a major target site for this cancer preventive agent.
Sigma-Aldrich
MRP4 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP5 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP7 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP1 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
Caco-2 Engineered Control Cells, one assay ready, 24-well plate
Sigma-Aldrich
Caco-2 Engineered Control Cells, one vial of frozen cells
Sigma-Aldrich
MDR1 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MDR1/MRP2 Double Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MDR1/BCRP Double Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP2 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
BCRP/MRP2 Double Knockout Caco-2 Cells, one vial
Sigma-Aldrich
BCRP Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP3 Knockout Caco-2 Cells, one vial