Merck
  • Diet-induced (epigenetic) changes in bone marrow augment atherosclerosis.
Diet-induced (epigenetic) changes in bone marrow augment atherosclerosis.
Journal of leukocyte biology (2014-07-16)
Erik van Kampen, Armand Jaminon, Theo J C van Berkel, Miranda Van Eck
ABSTRACT
Alterations in DNA methylation patterns in peripheral blood leukocytes precede atherosclerotic lesion development in mouse models of atherosclerosis and have been linked to cardiovascular death in patients. The aim of this study is to investigate the long-term changes induced by WTD feeding on BM cells and the consequences for atherosclerosis susceptibility. Hereto, WTD BM or Chow BM was transplanted into LDLR KO mice on chow. BM from WTD BM recipient mice exhibited hypomethylation of CpG regions in the genes encoding Pu.1 and IRF8, key regulators of monocyte proliferation and macrophage differentiation. In agreement, in blood, the numbers of leukocytes were 40% (P<0.05) higher as a result of an increase in F4/80(+) monocytes (3.4-fold; P<0.01). An increase of CD11c(++) cells was also found (2.4-fold; P<0.05). Furthermore, spleens were enlarged, and the percentage of F4/80(+) cells expressing CD86 was induced (1.8-fold; P<0.01), indicating increased activation of splenic macrophages. Importantly, mice reconstituted with WTD BM showed a significant, 1.4-fold (P<0.05) increase in aortic root plaque size in the absence of changes in serum cholesterol. We conclude that WTD challenge induces transplantable epigenetic changes in BM, alterations in the hematopoietic system, and increased susceptibility to atherosclerosis. Manipulation of the epigenome, when used in conjunction with blood lipid reduction, could thus prove beneficial to treat cardiovascular disorders.
Product Description
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sitaxentan sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture