Merck
  • A fine balance: Regulation of hippocampal Arc/Arg3.1 transcription, translation and degradation in a rat model of normal cognitive aging.
A fine balance: Regulation of hippocampal Arc/Arg3.1 transcription, translation and degradation in a rat model of normal cognitive aging.
Neurobiology of learning and memory (2014-08-26)
Bonnie R Fletcher, Gordon S Hill, Jeffrey M Long, Michela Gallagher, Matthew L Shapiro, Peter R Rapp
ABSTRACT
Memory decline is a common feature of aging. Expression of the immediate-early gene Arc is necessary for normal long-term memory, and although experience dependent Arc transcription is reportedly reduced in the aged rat hippocampus, it has not been clear whether this effect is an invariant consequence of growing older, or a finding linked specifically to age-related memory impairment. Here we show that experience dependent Arc mRNA expression in the hippocampus fails selectively among aged rats with spatial memory deficits. While these findings are consistent with the possibility that blunted Arc transcription contributes to cognitive aging, we also found increased basal ARC protein levels in the CA1 field of the hippocampus in aged rats with memory impairment, together with a loss of the experience dependent increase observed in young and unimpaired aged rats. Follow-up analysis revealed that increased basal translation and blunted ubiquitin mediated degradation may contribute to increased basal ARC protein levels noted in memory impaired aged rats. These findings indicate that Arc expression is regulated at multiple levels, and that several of these mechanisms are altered in cognitively impaired aged rats. Defining the influence of these alterations on the spatial and temporal fidelity of synapse specific, memory-related plasticity in the aged hippocampus is an important challenge.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Anti-Upf1 Antibody, from rabbit
SAFC
BIS-TRIS
SAFC
BIS-TRIS