Merck
  • Preparation and evaluation of ritodrine buccal tablets for rational therapeutic use.
Preparation and evaluation of ritodrine buccal tablets for rational therapeutic use.
International journal of pharmaceutics (2014-04-09)
Hiraku Onishi, Kei Yumoto, Osamu Sakata
ABSTRACT
Ritodrine hydrochloride (RD-HCl) tablets containing alginate (AL) and lactose (LC) with or without microcrystalline cellulose (MC) as excipients were produced as a buccal dosage form. The RD-HCl (2 mg) tablets with AL/LC but no MC swelled and dissolved gradually in the in vitro dissolution test. The tablet showing the fastest dissolution and highest drug release rate, called Tablet A1, was selected as a tablet to show rapid and prolonged absorption. However, in the in vivo buccal absorption test using rats, it could not give a plasma concentration over the human minimal effective level (15 ng/mL). The modified tablet containing AL, LC, MC and RD-HCl (4 mg), named Tablet B/MC, showed better hardness and faster drug release. Tablet B/MC gave a plasma concentration over the human effective level within 15 min, and the plasma concentration was maintained at >15 ng/mL over 4 h. Moreover, the deconvolution analyses demonstrated that a prolonged high absorption rate could be achieved in vivo best with Tablet B/MC. Tablet B/MC improved the pharmacokinetic profile in comparison with Tablet A1 and the solution dosage form. The RD-HCl buccal tablets with AL, LC and MC as excipients are suggested to be possibly useful for the treatment of premature labor.
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Supelco
Urethane, analytical standard
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Ritodrine hydrochloride
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O