Merck
A general and scalable synthesis approach to porous graphene.
Nature communications (2014-09-03)
Ding Zhou, Yi Cui, Pei-Wen Xiao, Mei-Yang Jiang, Bao-Hang Han
ABSTRACT
Porous graphene, which features nano-scaled pores on the sheets, is mostly investigated by computational studies. The pores on the graphene sheets may contribute to the improved mass transfer and may show potential applications in many fields. To date, the preparation of porous graphene includes chemical bottom-up approach via the aryl-aryl coupling reaction and physical preparation by high-energy techniques, and is generally conducted on substrates with limited yields. Here we show a general and scalable synthesis method for porous graphene that is developed through the carbothermal reaction between graphene and metal oxide nanoparticles produced from oxometalates or polyoxometalates. The pore formation process is observed in situ with the assistance of an electron beam. Pore engineering on graphene is conducted by controlling the pore size and/or the nitrogen doping on the porous graphene sheets by varying the amount of the oxometalates or polyoxometalates, or using ammonium-containing oxometalates or polyoxometalates.
Sigma-Aldrich
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 99.98% trace metals basis
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Molybdenum(IV) oxide, 99%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil