Merck
  • Fumigant toxicity and acetylcholinesterase inhibitory activity of 4 Asteraceae plant essential oils and their constituents against Japanese termite (Reticulitermes speratus Kolbe).
Fumigant toxicity and acetylcholinesterase inhibitory activity of 4 Asteraceae plant essential oils and their constituents against Japanese termite (Reticulitermes speratus Kolbe).
Pesticide biochemistry and physiology (2014-07-24)
Seon-Mi Seo, Junheon Kim, Jaesoon Kang, Sang-Hyun Koh, Young-Joon Ahn, Kyu-Suk Kang, Il-Kwon Park
ABSTRACT
This study investigated the fumigant toxicity of 4 Asteraceae plant essential oils and their constituents against the Japanese termite Reticulitermes speratus Kolbe. Fumigant toxicity varied with plant essential oils or constituents, exposure time, and concentration. Among the tested essential oils, those from Chamaemelum nobile exhibited the strongest fumigant toxicity, followed by those from Santolina chamaecyparissus, Ormenis multicaulis, and Eriocephalus punctulatus at 2 days after treatment. In all, 15, 24, 19, and 9 compounds were identified in the essential oils from C. nobile, E. punctulatus, O. multicaulis, and S. chamaecyparissus, respectively, by using gas chromatography, gas chromatography-mass spectrometry, or open-column chromatography. The identified compounds were tested individually for their fumigant toxicity against Japanese termites. Among the test compounds, trans-pinocarveol, caryophyllene oxide, sabinene hydrate, and santolina alcohol showed strong fumigant toxicity against Japanese termites. Acetylcholinesterase (AChE) inhibition activity of the identified compounds from C. nobile, E. punctulatus, O. multicaulis, and S. chamaecyparissus essential oils were tested to determine the mode of their action. The IC50 values of (+)-α-pinene, (-)-limonene, (-)-α-pinene, β-pinene, and β-phellandrene against Japanese termite AChE were 0.03, 0.13, 0.41, 0.42, and 0.67mg/mL, respectively. Further studies are warranted to determine the potential of these essential oils and their constituents as fumigants for termite control.
Product Description
Supelco
Linalool, analytical standard
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
Terpinolene, ≥95%, FG
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
p-Cymene, 99%
Sigma-Aldrich
Myrcene, stabilized, FCC, FG
Supelco
(−)-Terpinen-4-ol, analytical standard
Supelco
Linalyl acetate, analytical standard
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Camphor, 96%
Sigma-Aldrich
p-Cymene, ≥97%, FG
Supelco
2-Methyl-1-propanol, analytical standard
Supelco
Eucalyptol, analytical standard
Supelco
p-Cymene, analytical standard
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Terpinolene, technical, ≥85% (GC)
Sigma-Aldrich
3-Methyl-2-buten-1-ol, ≥98%, FG
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%
Sigma-Aldrich
2-Methyl-1-propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Sigma-Aldrich
3,3-Dimethylacryloyl chloride, 97%, contains 400 ppm phenothiazine as inhibitor
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Supelco
(−)-Camphor, analytical standard
Supelco
Myrcene, analytical standard
Supelco
(+)-Terpinen-4-ol, analytical standard
Sigma-Aldrich
(±)-Camphor, ≥95.5%