Merck
  • Protein arginine N-methyltransferase substrate preferences for different nη-substituted arginyl peptides.
Protein arginine N-methyltransferase substrate preferences for different nη-substituted arginyl peptides.
Chembiochem : a European journal of chemical biology (2014-07-22)
Dylan Thomas, Timo Koopmans, Ted M Lakowski, Helmi Kreinin, Mynol I Vhuiyan, Shona A Sedlock, Jennifer M Bui, Nathaniel I Martin, Adam Frankel
ABSTRACT
Protein arginine N-methyltransferases (PRMTs) catalyze methyl-group transfer from S-adenosyl-L-methionine onto arginine residues in proteins. In this study, modifications were introduced at the guanidine moiety of a peptidyl arginine residue to investigate how changes to the PRMT substrate can modulate enzyme activity. We found that peptides bearing Nη-hydroxy or Nη-amino substituted arginine showed higher apparent kcat values than for the monomethylated substrate when using PRMT1, whereas this catalytic preference was not observed for PRMT4 and PRMT6. Methylation by compromised PRMT1 variants E153Q and D51N further supports the finding that the N-hydroxy substitution facilitates methyl transfer by tuning the reactivity of the guanidine moiety. In contrast, Nη-nitro and Nη-canavanine substituted substrates inhibit PRMT activity. These findings demonstrate that methylation of these PRMT substrates is dependent on the nature of the modification at the guanidine moiety.
Product Description
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Methionine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O