Merck
  • Antioxidant activity of caffeoyl-prolyl-histidine amide and its effects on PDGF-induced proliferation of vascular smooth muscle cells.
Antioxidant activity of caffeoyl-prolyl-histidine amide and its effects on PDGF-induced proliferation of vascular smooth muscle cells.
Amino acids (2014-09-15)
Seon-Yeong Kwak, Hyun Jung Lee, Jin-Kyoung Yang, Eun Jig Lee, MiRan Seo, Yoon-Sik Lee
ABSTRACT
Caffeic acid (CA) is one of the antioxidants found in plants, which protects vascular cells against vascular injuries from oxidative stress. In our previous study, caffeoyl-prolyl-histidine amide (CA-L-Pro-L-His-NH2; CA-PH; a CA derivative) was synthesized, which exhibited a strong antioxidant activity with sufficient stability. In this study, we investigated the role of CA-PH in vascular smooth muscle cells (VSMCs) and confirmed the enhanced antioxidant activity of CA-PH compared with that of CA. In in vitro tube assays, CA-PH showed a higher free-radical-scavenging activity and lipid-peroxidation-inhibition activity than those of CA. In VSMCs, CA-PH significantly reduced hydrogen peroxide-induced ROS generation and increased the expression of heme oxygenase-1. Moreover, CA-PH effectively inhibited the platelet-derived growth factor-induced cellular proliferation of VSMCs, which was confirmed by a decrease in the expression of the proliferating cell nuclear antigen and the phosphorylation of Akt.
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Fmoc-D-Pro-OH, ≥98.0%
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Fmoc-D-His(Trt)-OH, 97% (HPLC)
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Ammonium thiocyanate, powder
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
Ammonium thiocyanate, ACS reagent, ≥97.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing