Merck
  • The importance of formulation in the successful lyophilization of influenza reference materials.
The importance of formulation in the successful lyophilization of influenza reference materials.
Biologicals : journal of the International Association of Biological Standardization (2015-01-24)
Chinwe Duru, Carolyn Swann, Una Dunleavy, Barbara Mulloy, Paul Matejtschuk
ABSTRACT
Lyophilized Influenza antigen reference reagents are a critical resource in the quality control of influenza vaccines. A standard formulation has been used successfully at NIBSC for many years however, following the unexpected occurrence of a collapsed appearance in a particular batch a study was carried out to establish the impact of the sugar concentration in the formulation using modulated differential scanning calorimetry (mDSC) and nuclear magnetic resonance spectroscopy (NMR). There was a correlation between the presence and size of the mDSC eutectic temperature events and the freeze dried appearance of the cakes, which became progressively worse with increasing amounts of sugar. NMR spectroscopy could be used to positively identify and quantify the sugars in the formulations. MDSC can rapidly predict if the freeze dried appearance will be acceptable so as to assure the successful lyophilization of influenza reference preparations.
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture