Merck
  • Developing an ultra non-fouling SU-8 and PDMS hybrid microfluidic device by poly(amidoamine) engraftment.
Developing an ultra non-fouling SU-8 and PDMS hybrid microfluidic device by poly(amidoamine) engraftment.
Colloids and surfaces. B, Biointerfaces (2015-02-18)
Yubo Qin, Poying Yeh, Xingkai Hao, Xudong Cao
ABSTRACT
A new strategy to prepare an ultra non-fouling SU-8/poly(dimethylsiloxane) (PDMS) hybrid microfluidic system is presented. We report a simple method to bond both SU-8 and PDMS surfaces to prepare the hybrid microfluidic device and then simultaneously modify both SU-8 and PDMS surfaces using poly(amidoamine) (PAMAM), a highly hydrophilic dendrimer to enhance non-fouling properties of the hybrid microfluidic channel. Water contact angle, atomic force microscopy (AFM) and X-ray photoelectron spectroscopy (XPS) are used to characterize and confirm surface modifications. Finally the non-fouling performance of the resulting microfluidic system is tested using both microbeads and Escherichia coli O157:H7 bacterial cells. We demonstrate that the obtained hybrid microfluidic system shows a significant microbead adsorption suppression of 99.7%, an ultra non-fouling performance that has not been reported in the literature before; in addition, 95% bacteria surface adsorption suppression is also obtained. Considering the significantly improved non-fouling performance and the ease of preparing the hybrid microfluidic system, we anticipate that this strategy will find wide applications in biosensing and detection.
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Supelco
Toluene, analytical standard
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
PAMAM dendrimer, ethylenediamine core, generation 7.0 solution, 5 wt. % in methanol
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%