Merck
Identification of natural indigo in historical textiles by GC-MS.
Analytical and bioanalytical chemistry (2015-01-19)
Laura Degani, Chiara Riedo, Oscar Chiantore
ABSTRACT
The possibility of successfully applying a common GC-MS procedure for identification in one step of all types of dyes from plants of unknown origin and from historical objects is particularly attractive due to the high separation efficiency of the capillary columns, the MS detection sensitivity and the reproducibility of results. In this work, GC-MS analysis, previously and successfully used for the characterization of anthraquinones, flavonoids and tannins from plant extracts and historical samples, has been tested on indigoid dyestuffs. An analytical procedure based on the silylating agent N,O-bis-(trimethylsilyl)trifluoroacetamide (BSTFA) with 1% trimethylchlorosilane (TMCS) was applied to pure molecules of indigotin and indirubin and to plant extracts of Indigofera tinctoria L. and Isatis tinctoria L. Preliminary tests have been done to establish the chromatographic conditions and the derivatization amounts most suitable for the simultaneous detection of indigoid molecules and of the other natural compounds, such as fatty acids, carboxylic acids and sugars, contained within the plant extracts. In order to assess the capacity and the sensitivity of the analytical procedure in typical archaeometric applications, wool samples dyed in the laboratory with indigo were analysed by mimicking the sample amounts typically available with historical objects. The electron ionization (EI) spectra of the main silylated derivatives of indigoid molecules obtained in this way constitute the necessary data set for the characterization of natural extracts and historical works of art. Subsequently, the procedure has been applied to historical samples for the detection of indigo and of other dyestuffs eventually contained in samples. Additional information, useful for restoration and preservation of works of art, could be also obtained on the nature of stains and smudges present on the sampled textile material. The GC-MS method turns out to be an efficient and fast analytical tool also for the identification of natural indigo in plants and textile artefacts, providing results complementary to those from high-performance liquid chromatography (HPLC).
Product Description
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Metolachlor OA, PESTANAL®, analytical standard
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥99.0%