Merck
  • Hapten synthesis, monoclonal antibody production and development of a competitive indirect enzyme-linked immunosorbent assay for erythromycin in milk.
Hapten synthesis, monoclonal antibody production and development of a competitive indirect enzyme-linked immunosorbent assay for erythromycin in milk.
Food chemistry (2014-10-14)
Zhanhui Wang, Tiejun Mi, Ross C Beier, Huiyan Zhang, Yajie Sheng, Weimin Shi, Suxia Zhang, Jianzhong Shen
ABSTRACT
Erythromycin is an antibiotic used extensively in veterinary practice worldwide for treatment, prevention and growth promotion. In this work, monoclonal antibodies (Mabs) against erythromycin were produced and used to develop a competitive indirect enzyme-linked immunosorbent assay (ciELISA) for the determination of erythromycin in milk. A novel carboxyphenyl derivative of erythromycin (ERO-CMO) was synthesized and conjugated with bovine serum (BSA) for use as the immunogen or ovalbumin (OVA) as the coating antigen. Four hybridoma cell lines were isolated, which produced Mabs that competed with erythromycin. The 6C1 and 5B2 Mabs had IC50 values for erythromycin of 14.40 and 0.94 μg L(-)(1), respectively. These Mabs demonstrated high cross-reactivity to the macrolides containing 14-membered rings, but not to oleandomycin. No cross-reactivity was observed for 12 macrolides that contained 15 or 16-membered lactone rings or for 2 pleuromutilins. The ciELISA developed using the 5B2 Mab afforded recovery values that ranged from 76.9% to 85.7% with only a 10-fold sample dilution prior to analysis.
Sigma-Aldrich
Tributylamine, puriss. plus, ≥99.5% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Tributylamine, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Isobutyl chloroformate, 98%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Erythromycin ethyl succinate, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin ethyl succinate, United States Pharmacopeia (USP) Reference Standard
USP
Desosaminylazithromycin, United States Pharmacopeia (USP) Reference Standard
USP
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tributylamine, ≥98.5%
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Roxithromycin, European Pharmacopoeia (EP) Reference Standard
Josamycin, European Pharmacopoeia (EP) Reference Standard
Azithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard