Merck
  • PDADMAC flocculation of Chinese hamster ovary cells: enabling a centrifuge-less harvest process for monoclonal antibodies.
PDADMAC flocculation of Chinese hamster ovary cells: enabling a centrifuge-less harvest process for monoclonal antibodies.
mAbs (2015-02-24)
Thomas McNerney, Anne Thomas, Anna Senczuk, Krista Petty, Xiaoyang Zhao, Rob Piper, Juliane Carvalho, Matthew Hammond, Satin Sawant, Jeanine Bussiere
ABSTRACT
High titer (>10 g/L) monoclonal antibody (mAb) cell culture processes are typically achieved by maintaining high viable cell densities over longer culture durations. A corresponding increase in the solids and sub-micron cellular debris particle levels are also observed. This higher burden of solids (≥15%) and sub-micron particles typically exceeds the capabilities of a continuous centrifuge to effectively remove the solids without a substantial loss of product and/or the capacity of the harvest filtration train (depth filter followed by membrane filter) used to clarify the centrate. We discuss here the use of a novel and simple two-polymer flocculation method used to harvest mAb from high cell mass cell culture processes. The addition of the polycationic polymer, poly diallyldimethylammonium chloride (PDADMAC) to the cell culture broth flocculates negatively-charged cells and cellular debris via an ionic interaction mechanism. Incorporation of a non-ionic polymer such as polyethylene glycol (PEG) into the PDADMAC flocculation results in larger flocculated particles with faster settling rate compared to PDADMAC-only flocculation. PDADMAC also flocculates the negatively-charged sub-micron particles to produce a feed stream with a significantly higher harvest filter train throughput compared to a typical centrifuged harvest feed stream. Cell culture process variability such as lactate production, cellular debris and cellular densities were investigated to determine the effect on flocculation. Since PDADMAC is cytotoxic, purification process clearance and toxicity assessment were performed.
Sigma-Aldrich
Sodium L-lactate, ≥99.0% (NT)
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Saponin from Quillaja sp., Sapogenin content 20-35 %
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Diallyldimethylammonium chloride solution, 65 wt. % in H2O
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 3,000
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 1,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 1,500
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, for molecular biology, 6,000
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic