Merck
  • Differential contribution of monocytes to heart macrophages in steady-state and after myocardial infarction.
Differential contribution of monocytes to heart macrophages in steady-state and after myocardial infarction.
Circulation research (2014-05-03)
Timo Heidt, Gabriel Courties, Partha Dutta, Hendrik B Sager, Matt Sebas, Yoshiko Iwamoto, Yuan Sun, Nicolas Da Silva, Peter Panizzi, Anja M van der Laan, Anja M van der Lahn, Filip K Swirski, Ralph Weissleder, Matthias Nahrendorf
ABSTRACT
Macrophages populate the steady-state myocardium. Previously, all macrophages were thought to arise from monocytes; however, it emerged that, in several organs, tissue-resident macrophages may self-maintain through local proliferation. Our aim was to study the contribution of monocytes to cardiac-resident macrophages in steady state, after macrophage depletion in CD11b(DTR/+) mice and in myocardial infarction. Using in vivo fate mapping and flow cytometry, we estimated that during steady state the heart macrophage population turns over in ≈1 month. To explore the source of cardiac-resident macrophages, we joined the circulation of mice using parabiosis. After 6 weeks, we observed blood monocyte chimerism of 35.3±3.4%, whereas heart macrophages showed a much lower chimerism of 2.7±0.5% (P<0.01). Macrophages self-renewed locally through proliferation: 2.1±0.3% incorporated bromodeoxyuridine 2 hours after a single injection, and 13.7±1.4% heart macrophages stained positive for the cell cycle marker Ki-67. The cells likely participate in defense against infection, because we found them to ingest fluorescently labeled bacteria. In ischemic myocardium, we observed that tissue-resident macrophages died locally, whereas some also migrated to hematopoietic organs. If the steady state was perturbed by coronary ligation or diphtheria toxin-induced macrophage depletion in CD11b(DTR/+) mice, blood monocytes replenished heart macrophages. However, in the chronic phase after myocardial infarction, macrophages residing in the infarct were again independent from the blood monocyte pool, returning to the steady-state situation. In this study, we show differential contribution of monocytes to heart macrophages during steady state, after macrophage depletion or in the acute and chronic phase after myocardial infarction. We found that macrophages participate in the immunosurveillance of myocardial tissue. These data correspond with previous studies on tissue-resident macrophages and raise important questions on the fate and function of macrophages during the development of heart failure.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard