Merck
  • Ligand-based virtual screening identifies a family of selective cannabinoid receptor 2 agonists.
Ligand-based virtual screening identifies a family of selective cannabinoid receptor 2 agonists.
Bioorganic & medicinal chemistry (2014-12-10)
Matteo Gianella-Borradori, Ivy Christou, Carole J R Bataille, Rebecca L Cross, Graham M Wynne, David R Greaves, Angela J Russell
ABSTRACT
The cannabinoid receptor 2 (CB2R) has been linked with the regulation of inflammation, and selective receptor activation has been proposed as a target for the treatment of a range of inflammatory diseases such as atherosclerosis and arthritis. In order to identify selective CB2R agonists with appropriate physicochemical and ADME properties for future evaluation in vivo, we first performed a ligand-based virtual screen. Subsequent medicinal chemistry optimisation studies led to the identification of a new class of selective CB2R agonists. Several examples showed high levels of activity (EC50<200 nM) and binding affinity (Ki<200 nM) for the CB2R, and no detectable activity at the CB1R. The most promising example, DIAS2, also showed favourable in vitro metabolic stability and absorption properties along with a clean selectivity profile when evaluated against a panel of GPCRs and kinases.
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)
Sigma-Aldrich
Pentyl acetate, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pentyl acetate, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%