Merck
  • Direct tandem mass spectrometric profiling of sulfatides in dry urinary samples for screening of metachromatic leukodystrophy.
Direct tandem mass spectrometric profiling of sulfatides in dry urinary samples for screening of metachromatic leukodystrophy.
Clinica chimica acta; international journal of clinical chemistry (2013-07-11)
Ladislav Kuchař, Befekadu Asfaw, Helena Poupětová, Jitka Honzíková, František Tureček, Jana Ledvinová
ABSTRACT
Prediagnostic steps in suspected metachromatic leukodystrophy (MLD) rely on clinical chemical methods other than enzyme assays. We report a new diagnostic method which evaluates changes in the spectrum of molecular types of sulfatides (3-O-sulfogalactosyl ceramides) in MLD urine. The procedure allows isolation of urinary sulfatides by solid-phase extraction on DEAE-cellulose membranes, transportation of a dry membrane followed by elution and tandem mass spectrometry (MS/MS) analysis in the clinical laboratory. Major sulfatide isoforms are normalized to the least variable component of the spectrum, which is the indigenous C18:0 isoform. This procedure does not require the use of specific internal standards and minimizes errors caused by sample preparation and measurement. Urinary sulfatides were analyzed in a set of 21 samples from patients affected by sulfatidosis. The combined abundance of the five most elevated isoforms, C22:0, C22:0-OH, C24:0, C24:1-OH, and C24:0-OH sulfatides, was found to give the greatest distinction between MLD-affected patients and a control group. The method avoids transportation of liquid urine samples and generates stable membrane-bound sulfatide samples that can be stored at ambient temperature. MS/MS sulfatide profiling targeted on the most MLD-representative isoforms is simple with robust results and is suitable for screening.
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer