Merck
  • ZFC3H1, a zinc finger protein, modulates IL-8 transcription by binding with celastramycin A, a potential immune suppressor.
ZFC3H1, a zinc finger protein, modulates IL-8 transcription by binding with celastramycin A, a potential immune suppressor.
PloS one (2014-10-01)
Takeshi Tomita, Katsuaki Ieguchi, Frédéric Coin, Fredric Coin, Yasuhiro Kato, Haruhisa Kikuchi, Yoshiteru Oshima, Shoichiro Kurata, Yoshiro Maru
ABSTRACT
Celastramycin A, a small molecule that inhibits the production of antibacterial peptides in an ex vivo culture system of Drosophila, suppresses the TNFα-mediated induction of IL-8 in mammalian cells. To understand its molecular mechanism, we examined Celastramycin A binding proteins and investigated their biological functions. Our screening and subsequent pull-down assay revealed ZFC3H1 (also known as CCDC131 or CSRC2), an uncharacterized zinc finger protein, as a Celastramycin A binding protein. The knockdown of ZFC3H1 reduced IL-8 expression levels in the TNFα-stimulated lung carcinoma cell line, LU99, and UV-irradiated HeLa cells. Based on reporter assay results, we concluded that ZFC3H1 participates in the transcriptional activation of IL-8. The findings of our UV-irradiation experiments implied that ZFC3H1 may indirectly interact with ERCC1 in an activated DNA repair complex. Thus, we designated ZFC3H1 as a mammalian target of Celastramycin A (mTOC).
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SAFC
HEPES