Merck
  • Dorsomedial hypothalamus CRF type 1 receptors selectively modulate inhibitory avoidance responses in the elevated T-maze.
Dorsomedial hypothalamus CRF type 1 receptors selectively modulate inhibitory avoidance responses in the elevated T-maze.
Behavioural brain research (2014-06-18)
Mariana S C F Silva, Bruno A Pereira, Isabel C Céspedes, Juliana O G Nascimento, Jackson C Bittencourt, Milena B Viana
ABSTRACT
Corticotropin-releasing factor (CRF) plays a critical role in the mediation of physiological and behavioral responses to stressors. In the present study, we investigated the role played by the CRF system within the dorsomedial hypothalamus (DMH) in the modulation of anxiety- and panic-related responses. Male Wistar rats were administered into the DMH with CRF (125 and 250 ng/0.2 μl, experiment 1) or with the CRFR1 antagonist antalarmin (25 ng/0.2 μl, experiment 2) and 10 min later tested in the elevated T-maze (ETM) for inhibitory avoidance and escape measurements. In clinical terms, these responses have been respectively related to generalized anxiety and panic disorder. To further verify if the anxiogenic effects of CRF were mediated by CRFR1 activation, we also investigated the effects of the combined treatment with CRF (250 ng/0.2 μl) and antalarmin (25 ng/0.2 μl) (experiment 3). All animals were tested in an open field, immediately after the ETM, for locomotor activity assessment. Results showed that 250 ng/0.2μl of CRF facilitated ETM avoidance, an anxiogenic response. Antalarmin significantly decreased avoidance latencies, an anxiolytic effect, and was able to counteract the anxiogenic effects of CRF. None of the compounds administered altered escape responses or locomotor activity measurements. These results suggest that CRF in the DMH exerts anxiogenic effects by activating type 1 receptors, which might be of relevance to the physiopathology of generalized anxiety disorder.
Sigma-Aldrich
Flunixin meglumine, ≥98% (HPLC)
Sigma-Aldrich
Antalarmin hydrochloride, ≥98% (HPLC), solid
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
Supelco
Flunixin Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material