Merck
  • The influence of linker length on the properties of cathepsin S cleavable (177)Lu-labeled HPMA copolymers for pancreatic cancer imaging.
The influence of linker length on the properties of cathepsin S cleavable (177)Lu-labeled HPMA copolymers for pancreatic cancer imaging.
Biomaterials (2014-04-24)
Wen Shi, Sunny M Ogbomo, Nilesh K Wagh, Zhengyuan Zhou, Yinnong Jia, Susan K Brusnahan, Jered C Garrison
ABSTRACT
N-(2-hydroxypropyl)-methacrylamide (HPMA) copolymers have shown promise for application in the detection and staging of cancer. However, non-target accumulation, particularly in the liver and spleen, hinders the detection of resident or nearby metastatic lesions thereby decreasing diagnostic effectiveness. Our laboratory has pursued the development of cathepsin S susceptible linkers (CSLs) to reduce the non-target accumulation of diagnostic/radiotherapeutic HPMA copolymers. In this study, we ascertain if the length of the linking group impacts the cleavage and clearance kinetics, relative to each other and a non-cleavable control, due to a reduction in steric inhibition. Three different CSLs with linking groups of various lengths (0, 6 and 13 atoms) were conjugated to HPMA copolymers. In vitro cleavage studies revealed that the longest linking group (13 atoms) led to more rapid cleavage when challenged with cathepsin S. The CSL incorporated HPMA copolymers demonstrated significantly higher levels of excretion and a significant decrease in long-term hepatic and splenic retention relative to the non-cleavable control. Contrary to in vitro observations, the length of the linking group did not substantially impact the non-target in vivo clearance. In the case of HPAC tumor retention, the CSL with the null (0 atom) linker demonstrated significantly higher levels of retention relative to the other CSLs. Given these results, we find that the length of the linking group of the CSLs did not substantially impact non-target clearance, but did influence tumor retention. Overall, these results demonstrate that the CSLs can substantially improve the non-target clearance of HPMA copolymers thereby enhancing clinical potential.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
N-(2-Hydroxypropyl)-2-methyl-prop-2-enamide, AldrichCPR
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
4,4′-Azobis(4-cyanovaleric acid), ≥98.0% (T)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
1-Methyl-2-pyrrolidinone, analytical standard