Merck
  • Actions of methyl-, propyl- and butylparaben on estrogen receptor-α and -β and the progesterone receptor in MCF-7 cancer cells and non-cancerous MCF-10A cells.
Actions of methyl-, propyl- and butylparaben on estrogen receptor-α and -β and the progesterone receptor in MCF-7 cancer cells and non-cancerous MCF-10A cells.
Toxicology letters (2014-08-17)
Anna Maria Wróbel, Ewa Łucja Gregoraszczuk
ABSTRACT
Numerous studies have shown that widely used parabens possess estrogenic properties. In the present study, we examined the effects of methyl-, propyl- and butylparaben on the mRNA and protein expression of estrogen receptor (ER)-α (ESR1) and -β (ESR2) and the progesterone receptor (PGR). Human MCF-7 breast cancer cells and MCF-10A non-transformed breast epithelial cells were exposed to parabens at a concentration of 20nM; 17β-estradiol at a concentration of 10nM, was used as a positive control. Both propyl- and butylparaben stimulated PGR mRNA expression in MCF-7 cells, whereas methyl- and propylparaben PGR protein expression. In MCF-10A cells, butyl- and propylparaben increased only PGR mRNA expression. All parabens increased ESR1 gene and protein expression in MCF-7 and with the exception of butylparaben in MCF-10A cells. All parabens significantly increased ESR2 mRNA and protein expression in MCF-7 cells, but in MCF-10A cells only ESR2 protein expression. In summary, by virtue of their stimulatory action on the expression of ESR1, ESR2 and PGR in cancer cells, parabens can be viewed as potential contributors to breast cancer progression. Extension, the actions of these parabens on the expression of ERs and PGR in non-cancerous cells point to possible actions on breast cancer initiation.
Product Description
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Butyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propyl 4-hydroxybenzoate sodium salt, tested according to Ph. Eur.
USP
Methylparaben, United States Pharmacopeia (USP) Reference Standard
USP
Propylparaben, United States Pharmacopeia (USP) Reference Standard
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl 4-hydroxybenzoate sodium salt, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Estradiol, meets USP testing specifications
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
USP
Butylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC