Merck
  • Rh(III)-catalyzed selective coupling of N-methoxy-1H-indole-1-carboxamides and aryl boronic acids.
Rh(III)-catalyzed selective coupling of N-methoxy-1H-indole-1-carboxamides and aryl boronic acids.
Organic letters (2014-06-25)
Jing Zheng, Yan Zhang, Sunliang Cui
ABSTRACT
A Rh(III)-catalyzed selective coupling of N-methoxy-1H-indole-1-carboxamide and aryl boronic acids is reported. The coupling is mild and efficient toward diverse product formation, with selective C-C and C-C/C-N bond formation. Kinetic isotope effects studies were conducted to reveal a mechanism of C-H activation and electrophilic addition.
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Copper - O.F.H.C., foil, 5m coil, thickness 0.075mm, half hard, 99.95+%
Copper, wire reel, 500m, diameter 0.125mm, annealed, 99.9%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.05mm, hard, 99.95+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.05mm, hard, 99.95+%
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Copper - O.F.H.C., foil, 6mm disks, thickness 0.006mm, hard, 99.95+%
Copper - O.F.H.C., foil, 50m coil, thickness 0.05mm, coil width 10mm, annealed, 99.95+%
Copper - O.F.H.C., foil, 5m coil, thickness 0.125mm, hard, 99.95+%
Copper, wire reel, 100m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Copper, wire reel, 1m, diameter 0.01mm, hard, 99.99%
Copper, wire reel, 500m, diameter 0.25mm, annealed, 99.9%
Copper, wire reel, 50m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 10m, diameter 0.02mm, annealed, 99.99%
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper, wire reel, 5000m, diameter 0.04mm, annealed, 99.9%