Merck
General toxicology of tin and its organic compounds.
Neurotoxicology (1984-01-01)
M R Krigman, A P Silverman
ABSTRACT
The toxicology of tin is almost entirely the toxicology of the organic compounds of tin, for the metal itself and its inorganic compounds appear to be nearly harmless for practical purposes. Furthermore, the neurotoxicity of organotin is essentially that of trimethyltin and triethyltin.
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Tin, foil, light tested, 50x50mm, thickness 0.008mm, 99.75%
Tin, foil, 10mm disks, thickness 0.02mm, 99.75%
Tin, foil, 15mm disks, thickness 0.075mm, as rolled, 98.8%
Tin, foil, 1m coil, thickness 0.030mm, as rolled, 97.4%
Tin, foil, 10mm disks, thickness 0.008mm, 99.75%
Tin, foil, 10mm disks, thickness 0.009mm, 99.75%
Tin, foil, 1m coil, thickness 0.072mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 15mm disks, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 0.030mm, as rolled, 97.4%
Tin, foil, 1m coil, thickness 0.25mm, as rolled, 98.8%
Tin, foil, 25mm disks, thickness 0.0125mm, 97.4%
Tin, foil, 10mm disks, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 1m coil, thickness 0.075mm, as rolled, 98.8%
Tin, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Tin, foil, 25mm disks, thickness 0.025mm, as rolled, 99.99+%
Tin, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Tin, foil, 1m coil, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 1m coil, thickness 0.01mm, 97.4%
Tin, foil, 4mm disks, thickness 0.004mm, 99.75%
Tin, foil, 15mm disks, thickness 0.15mm, as rolled, 99.99+%
Tin, foil, 15mm disks, thickness 0.009mm, 97.4%
Tin, foil, 10mm disks, thickness 0.01mm, 99.75%
Tin, foil, 1m coil, thickness 0.125mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 0.0125mm, 99.75%