Merck
  • [Progress on tantalum rod implanting for the treatment of femur head necrosis].
[Progress on tantalum rod implanting for the treatment of femur head necrosis].
Zhongguo gu shang = China journal of orthopaedics and traumatology (2013-10-19)
Xiao-kang Tang, Fu-sheng Ye, Pei-jian Tong, Yan-hua Fan, Min Li, Hang Ying, Lu-wei Xiao
ABSTRACT
Incorrect treatment for femur head necrosis can cause collapse of femoral head and tresult in severe harm for the patients (especially for the patient with middle-aged and young). The structure and mechanics characteristics of tantalum rod is similar to bone tissue, it higher strength and can adapt the internal environment of organism, so it has a large potency in treating femur head necrosis. Treatment of early femur head necrosis with tantalum rod implanting had alreadly widey applied at home and abroad, the method has the advantages of simple operation, little risk, less complication and beseems the patient with stage I - II of ARCO. But reasons that the difficult diagnosis of early femur head necrosis, localized effect of tantalum rod, different experience of medical worker,caused the contentions about effect of tantalum rod implanting. With development of science, tantalum rod implanting combined with correlative biotechnology should raise the effect in treating femur head necrosis.
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Tantalum, foil, 10mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.20mm, annealed, 99.9%
Tantalum, foil, 6mm disks, thickness 0.02mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.25mm, annealed, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.02mm, 99.9%
Tantalum, foil, 4mm disks, thickness 0.5mm, annealed, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.015mm, 99.9%
Tantalum, mesh, 40x40mm, nominal aperture 0.43mm, wire diameter 0.075mm, 50x50 wires/inch, open area 72%, plain weave mesh
Tantalum, foil, 50mm disks, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, 8mm disks, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, light tested, 150x150mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, 5m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.0075mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.003mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.015mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.25mm, annealed, 99.99%
Tantalum, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, tube, 100mm, outside diameter 8.7mm, inside diameter 8.34mm, wall thickness 0.18mm, as drawn, 99.9%
Tantalum, pellets, 20g, max. size 10mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.005mm, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.004mm, 99.9%