Merck
Gentian violet: a 19th century drug re-emerges in the 21st century.
Experimental dermatology (2013-10-15)
Alexander M Maley, Jack L Arbiser
ABSTRACT
Gentian violet (GV) has a long and varied history as a medicinal agent. Historically used as an antibacterial and antifungal, recent reports have shown its utility as an antitypranosomal, antiviral and anti-angiogenic agent. The objective of this article is to summarize evidence regarding the efficacy and safety of GV use in dermatology. Recent discoveries have found novel targets of GV, namely NADPH oxidase in mammalian cells and thioredoxin reductase 2 in bacterial, fungal and parasitic cells. These discoveries have expanded the use of GV in the 21st century. Given that GV is well tolerated, effective and inexpensive, its use in dermatology is predicted to increase.
Sigma-Aldrich
Gram′s crystal violet solution, for microscopy
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard