Merck
The salicylic acid loop in plant defense.
Current opinion in plant biology (2003-07-23)
Jyoti Shah
ABSTRACT
Salicylic acid is an important signal molecule in plant defense. In the past two years, significant progress has been made in understanding the mechanism of salicylic-acid biosynthesis and signaling in plants. A pathway similar to that found in some bacteria synthesizes salicylic acid from chorismate via isochorismate. Salicylic-acid signaling is mediated by at least two mechanisms, one requiring the NON-EXPRESSOR OF PR1 (NPR1) gene and a second that is independent of NPR1. Feedback loops involving salicylic acid modulate upstream signals. These feedback loops may provide a point for integrating developmental, environmental and other defense-associated signals, and thus fine-tune the defense responses of plants.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%